accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOSOOH2N

CAS RN 902837-00-7

IUPAC Name: 4-(2-formylphenoxy)benzene-1-sulfonamide
Synonyms: 4-(2-formylphenoxy)benzenesulfonamide
Molecular Weight (g/mol): 277.29
Molecular Formula: C13H11NO4S
InChi Key: XPTCFGSAQWFPKM-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(OC2=CC=CC=C2C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results