Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Formylphenoxy)benzenesulfonamide, Thermo Scientific™

Catalog No. AAH51005MD
Change view
Click to view available options
Quantity:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51005MD 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH51005MD Supplier Thermo Scientific Chemicals Supplier No. H51005MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as intermediate for pharmaceutical and agrochemicals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Specifications

CAS 902837-00-7
Molecular Formula C13H11NO4S
Quantity 250mg
MDL Number MFCD08061110
Synonym 4-2-formylphenoxy benzenesulfonamide, 4-2-formyl-phenoxy-benzenesulfonamide, 4-2-formylphenoxy benzene-1-sulfonamide, benzenesulfonamide pound not4-2-formylphenoxy
InChI Key XPTCFGSAQWFPKM-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C=O)OC2=CC=C(C=C2)S(=O)(=O)N
IUPAC Name 4-(2-formylphenoxy)benzenesulfonamide
Molecular Weight (g/mol) 277.294
PubChem CID 42553490
Formula Weight 277.3
Chemical Name or Material 4-(2-Formylphenoxy)benzenesulfonamide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.