accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONI

CAS RN 902837-52-9

IUPAC Name: 2-ethoxy-5-iodopyridine
Synonyms: 2-ethoxy-5-iodopyridine
Molecular Weight (g/mol): 249.05
Molecular Formula: C7H8INO
InChi Key: AIFZXWONOPNLLM-UHFFFAOYSA-N
SMILES: CCOC1=NC=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results