accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(R)(R)OH3CH2N

CAS RN 90290-05-4

IUPAC Name: (2R,5R)-2,5-bis(methoxymethyl)pyrrolidin-1-ium
Synonyms: (2R,5R)-2,5-bis(methoxymethyl)pyrrolidin-1-ium
Molecular Weight (g/mol): 160.24
Molecular Formula: C8H18NO2
InChi Key: HDXYTIYVVNJFLU-HTQZYQBOSA-O
SMILES: COC[C@H]1CC[C@H](COC)[NH2+]1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)