Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R,R)-(-)-2,5-Bis(methoxymethyl)pyrrolidine, 96%, Thermo Scientific™

Catalog No. AC293452500
Change view
Click to view available options
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC293452500 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC293452500 Supplier Thermo Scientific Chemicals Supplier No. 293452500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 90290-05-4
CAS Min % 95.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C8H17NO2
Refractive Index 1.4470 to 1.4500 (20°C, 589nm)
Quantity 250mg
Specific Rotation -7° to -9° (20°C, 589nm) (c=3, ethanol)
Synonym 2r,5r-2,5-bis methoxymethyl pyrrolidine, r,r---2,5-bis methoxymethyl pyrrolidine, pyrrolidine, 2,5-bis methoxymethyl-, 2r,5r, 2r,5r-5-methoxymethyl pyrrolidin-2-yl methoxymethane, r,r---2,5-bis methoxymethyl-pyrrolidine, r,r-2,5-bis methoxymethyl pyrrolidine, 2beta,5alpha-bis methoxymethyl pyrrolidine
InChI Key HDXYTIYVVNJFLU-HTQZYQBOSA-N
SMILES COCC1CCC(N1)COC
IUPAC Name (2R,5R)-2,5-bis(methoxymethyl)pyrrolidine
Molecular Weight (g/mol) 159.229
PubChem CID 2724974
Formula Weight 159.23
Percent Purity ≥95%
Chemical Name or Material (R, R)-(-)-2, 5-Bis(methoxymethyl)pyrrolidine, 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.