accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONHSOO

CAS RN 90291-46-6

IUPAC Name: 2-(naphthalene-1-sulfonamido)-3-phenylpropanoic acid
Synonyms: 2-(naphthalene-1-sulfonamido)-3-phenylpropanoic acid
Molecular Weight (g/mol): 355.41
Molecular Formula: C19H17NO4S
InChi Key: NWHURKYFYGKGRT-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=CC=C1)NS(=O)(=O)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results