Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Naphthalenesulfonyl)-L-phenylalanine 98.0+%, TCI America™
SDP

Catalog No. N05795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N05795G Supplier TCI America Supplier No. N05795G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90291-46-6
Molecular Formula C19H17NO4S
Molecular Weight (g/mol) 355.408
MDL Number MFCD00191467
InChI Key NWHURKYFYGKGRT-KRWDZBQOSA-N
PubChem CID 22216410
IUPAC Name (2S)-2-(naphthalen-1-ylsulfonylamino)-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)O)NS(=O)(=O)C2=CC=CC3=CC=CC=C32

Specifications

Melting Point 141°C
Color White-Yellow
Quantity 5 g
Formula Weight 355.41
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(1-Naphthalenesulfonyl)-L-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.