CAS RN 9036-19-5
Igepal Ca - 630 Non-Ionic Detergent MP Biomedicals
CAS: 9036-19-5 Molecular Formula: (C2H4O)x·C14H22O Synonym: Polyethylene glycol octylphenol ether
Triton X-100™, Wetting Agent, LabChem™
CAS: 9036-19-5
Igepal(R) CA-720, Spectrum™ Chemical
CAS: 9036-19-5 Molecular Formula: C18H30O3 Molecular Weight (g/mol): 294.44 InChI Key: LBCZOTMMGHGTPH-UHFFFAOYSA-N IUPAC Name: 2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethan-1-ol SMILES: CC(C)(C)CC(C)(C)C1=CC=C(OCCOCCO)C=C1
Triton X-100, For Analysis, MilliporeSigma™
CAS: 9036-19-5 Molecular Formula: C8H17C6H4(OCH2CH2)nOH Synonym: Octoxynol
Triton™ X-114, MP Biomedicals™
CAS: 9036-19-5
Triton(R) X-405, 70% Solution in Water, Spectrum™ Chemical
CAS: 9036-19-5
Triton(R) X-100, Spectrum™ Chemical
CAS: 9036-19-5 Molecular Formula: C18H30O3 Molecular Weight (g/mol): 294.44 InChI Key: LBCZOTMMGHGTPH-UHFFFAOYSA-N IUPAC Name: 2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethan-1-ol SMILES: CC(C)(C)CC(C)(C)C1=CC=C(OCCOCCO)C=C1
Triton(R) X-100, Spectrum™ Chemical
CAS: 9036-19-5 Molecular Formula: C18H30O3 Molecular Weight (g/mol): 294.44 InChI Key: LBCZOTMMGHGTPH-UHFFFAOYSA-N IUPAC Name: 2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethan-1-ol SMILES: CC(C)(C)CC(C)(C)C1=CC=C(OCCOCCO)C=C1