Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Triton X-100, Protein Grade™ Detergent, 10% Solution, Sterile-Filtered, Calbiochem™,

Catalog No. 64846350ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-846-350ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 64-846-350ML Supplier MilliporeSigma™ Supplier No. 64846350ML
Add to Cart
Add to Cart

Specifications

Color Undesignated
Quantity 50 mL
Packaging Plastic Jar
Synonym Polyethylene Glycol-p-isooctylphenyl Ether
Grade Protein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.