accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90389-86-9

IUPAC Name: [(3-fluorophenyl)methyl](propyl)amine
Synonyms: [(3-fluorophenyl)methyl](propyl)amine
Molecular Weight (g/mol): 167.23
Molecular Formula: C10H14FN
InChi Key: OORHPLFSYCEZIP-UHFFFAOYSA-N
SMILES: CCCNCC1=CC=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results