accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90390-07-1

IUPAC Name: methyl({[3-(trifluoromethyl)phenyl]methyl})amine
Synonyms: methyl({[3-(trifluoromethyl)phenyl]methyl})amine
Molecular Weight (g/mol): 189.18
Molecular Formula: C9H10F3N
InChi Key: JFLPPELZYKHKQZ-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)