Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-[3-(trifluoromethyl)benzyl]amine, 97+%, Thermo Scientific™

Catalog No. MO00895DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO00895DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO00895DA Supplier Thermo Scientific Chemicals Supplier No. MO00895DA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90390-07-1
Molecular Formula C9H10F3N
Molecular Weight (g/mol) 189.181
InChI Key JFLPPELZYKHKQZ-UHFFFAOYSA-N
Synonym n-methyl-3-trifluoromethyl benzylamine, n-methyl-n-3-trifluoromethyl benzyl amine, n-methyl-1-3-trifluoromethyl phenyl methanamine, methyl 3-trifluoromethyl phenyl methyl amine, benzenemethanamine, n-methyl-3-trifluoromethyl, n-methyl 3-trifluoromethyl phenyl methanamine, n-methyl-1-3-trifluoromethyl phenyl methanamine hydrochloride, chembrdg-bb 4004318, methyl-3-trifluoromethyl-benzyl-amine
PubChem CID 485414
IUPAC Name N-methyl-1-[3-(trifluoromethyl)phenyl]methanamine
SMILES CNCC1=CC(=CC=C1)C(F)(F)F

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 189.18
Percent Purity 97%
Chemical Name or Material N-Methyl-N-[3-(trifluoromethyl)benzyl]amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.