accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOOOOOHCH3OOOHCH3OHOH3COOHH3COOHH3COOHCH3OOHCH3CH3OOOOCH3OOCH3OCH3OH3COH3COH3COCH3

CAS RN 9041-56-9

IUPAC Name: 1-{[3,4,5-tris(2-hydroxybutoxy)-6-{[4,5,6-tris(2-hydroxybutoxy)-2-[(2-hydroxybutoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}butan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
Synonyms: hydroxybutyl methylcellulose
Molecular Weight (g/mol): 1373.67
Molecular Formula: C64H124O30
InChi Key: RPZANUYHRMRTTE-UHFFFAOYNA-N
SMILES: COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CCC(O)COCC1OC(OC2C(COCC(O)CC)OC(OCC(O)CC)C(OCC(O)CC)C2OCC(O)CC)C(OCC(O)CC)C(OCC(O)CC)C1OCC(O)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results