Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxybutyl Methyl Cellulose, Thermo Scientific™

Catalog No. AC190651000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC190651000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC190651000 Supplier Thermo Scientific Chemicals Supplier No. 190651000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9041-56-9
Molecular Formula C64H124O30
Molecular Weight (g/mol) 1373.666
InChI Key RPZANUYHRMRTTE-UHFFFAOYSA-N
Synonym hydroxybutyl methyl cellulose, cellulose, hydroxybutyl methyl ether, hydroxybutyl methylcellulose, 2,3,4-trimethoxy-6-methoxymethyl-5-3,4,5-trimethoxy-6-methoxymethyl oxan-2-yl oxyoxane; 1-3,4,5-tris 2-hydroxybutoxy-6-4,5,6-tris 2-hydroxybutoxy-2-2-hydroxybutoxymethyl oxan-3-yl oxyoxan-2-yl methoxy butan-2-ol, 2,3,4-trimethoxy-6-methoxymethyl-5-3,4,5-trimethoxy-6-methoxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran; 1-3,4,5-tris 2-hydroxybutoxy-6-4,5,6-tris 2-hydroxybutoxy-2-2-hydroxybutoxymethyl tetrahydropyran-3-yl oxy-tetrahydropyran-2-yl methoxy
PubChem CID 3084047
IUPAC Name 2,3,4-trimethoxy-6-(methoxymethyl)-5-[3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane;1-[[3,4,5-tris(2-hydroxybutoxy)-6-[4,5,6-tris(2-hydroxybutoxy)-2-(2-hydroxybutoxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]butan-2-ol
SMILES CCC(COCC1C(C(C(C(O1)OC2C(OC(C(C2OCC(CC)O)OCC(CC)O)OCC(CC)O)COCC(CC)O)OCC(CC)O)OCC(CC)O)OCC(CC)O)O.COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Color White
Loss on Drying 10% max.
Infrared Spectrum Authentic
Quantity 100 g
Physical Form Powder
Chemical Name or Material Hydroxybutyl methyl cellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.