CAS RN 9041-92-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ α1-Antitrypsin, Human Plasma
α1-Antitrypsin, CAS 9041-92-3, is a protease inhibitor of the serpin superfamily. Inhibits elastase, cathepsin G, and proteinase 3.