Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ α1-Antitrypsin, Human Plasma

Catalog No. 1782515MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-825-15MG 5 mg
17-825-11MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-825-15MG Supplier MilliporeSigma™ Supplier No. 1782515MG
Add to Cart
Add to Cart

α1-Antitrypsin, CAS 9041-92-3, is a protease inhibitor of the serpin superfamily. Inhibits elastase, cathepsin G, and proteinase 3.

A single polypeptide chain of 394 amino acids and three carbohydrate side chains linked to asparagine residues present at about 290mg/100mL in acute phase plasma. Serine protease inhibitor that also acts as a major physiological regulator of elastase. Inhibitor of neutrophil elastase, cathepsin G, and proteinase 3.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 300 mM NaCl, 30 mM sodium phosphate buffer, pH 6.5.
Target Proteins serine proteases
CAS 9041-92-3
Formula Weight 52000g/mol
Solubility H2O
Form Solid
Quantity 5 mg
Purity 95% by SDS-PAGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.