accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90563-93-2

IUPAC Name: (2S)-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate
Synonyms: (2S)-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate
Molecular Weight (g/mol): 178.17
Molecular Formula: C9H8NO3
InChi Key: VNTNLTOJGXENIZ-QMMMGPOBSA-M
SMILES: [O-]C(=O)[C@@H]1CNC2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results