Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydro-2H-1,4-benzoxazine-2-carboxylic acid, ≥97%, Thermo Scientific™

Catalog No. CC29701CB
Change view
Click to view available options
Amber Glass Bottle; 250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC29701CB Amber Glass Bottle; 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC29701CB Supplier Thermo Scientific Chemicals Supplier No. CC29701CB

missing translation for 'validMainframeMsg'

Specifications

CAS 90563-93-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H9NO3
Quantity 250 mg
MDL Number MFCD05865108
Synonym 2h-1,4-benzoxazine-2-carboxylicacid, 3,4-dihydro, 2h-1,4-benzoxazine-2-carboxylicacid,3,4-dihydro, 2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro, 3,4-dihydro-2h-benzo b 1,4 oxazine-2-carboxylic acid, benzomorpholine-2-carboxylic acid, 2h,3h,4h-benzo e 1,4-oxazaperhydroine-2-carboxylic acid, +/--3,4-dihydro-2h-benzo b 1,4 oxazine-2-carboxylic acid
InChI Key VNTNLTOJGXENIZ-UHFFFAOYSA-N
SMILES C1C(OC2=CC=CC=C2N1)C(=O)O
IUPAC Name 3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid
Molecular Weight (g/mol) 179.175
PubChem CID 3826952
Formula Weight 179.17
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-1, 4-benzoxazine-2-carboxylic acid
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.