accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOFFFFF

CAS RN 906352-59-8

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 2-(morpholin-4-yl)benzoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 2-(morpholin-4-yl)benzoate
Molecular Weight (g/mol): 373.28
Molecular Formula: C17H12F5NO3
InChi Key: SMSUNMKSTOOWTF-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CC=CC=C2N2CCOCC2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results