Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorophenyl 2-morpholinobenzoate, 97%, Thermo Scientific™

Catalog No. CC24326CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC24326CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC24326CB Supplier Thermo Scientific Chemicals Supplier No. CC24326CB

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 906352-59-8
Molecular Formula C17H12F5NO3
Molecular Weight (g/mol) 373.28
MDL Number MFCD09025824
InChI Key SMSUNMKSTOOWTF-UHFFFAOYSA-N
Synonym pentafluorophenyl 2-morpholinobenzoate, pentafluorophenyl 2-morpholin-4-ylbenzoate, pentafluorophenyl 2-morpholin-4-yl benzoate, 2,3,4,5,6-pentafluorophenyl 2-morpholin-4-yl benzoate, perfluorophenyl 2-morpholinobenzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 2-morpholin-4-ylbenzoate, 2-4-morpholinyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229476
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CC=CC=C2N2CCOCC2)C(F)=C1F

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 2,3,4,5,6-pentafluorophenyl 2-(morpholin-4-yl)benzoate
Formula Weight 373.28
Percent Purity 95%
Chemical Name or Material Pentafluorophenyl 2-morpholinobenzoate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.