accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 906352-61-2

IUPAC Name: 2-[4-(chloromethyl)phenyl]-1,3-thiazole
Synonyms: 2-[4-(chloromethyl)phenyl]-1,3-thiazole
Molecular Weight (g/mol): 209.69
Molecular Formula: C10H8ClNS
InChi Key: BFAWMDHRKPPXSO-UHFFFAOYSA-N
SMILES: ClCC1=CC=C(C=C1)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results