Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[4-(Chloromethyl)phenyl]-1,3-thiazole, 97%, Thermo Scientific™

Catalog No. CC40224DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC40224DA 1 g Amber Glass Bottle
CC40224CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC40224DA Supplier Thermo Scientific Chemicals Supplier No. CC40224DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 906352-61-2
Molecular Formula C10H8ClNS
Molecular Weight (g/mol) 209.691
MDL Number MFCD09064951
InChI Key BFAWMDHRKPPXSO-UHFFFAOYSA-N
Synonym 2-4-chloromethyl phenyl-1,3-thiazole, 2-4-chloromethyl phenyl thiazole
PubChem CID 24229488
IUPAC Name 2-[4-(chloromethyl)phenyl]-1,3-thiazole
SMILES C1=CC(=CC=C1CCl)C2=NC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 209.7
Percent Purity 97%
Chemical Name or Material 2-[4-(Chloromethyl)phenyl]-1, 3-thiazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.