accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 906352-74-7

IUPAC Name: 1-{4-[5-(trifluoromethyl)pyridin-2-yl]phenyl}methanamine
Synonyms: 1-{4-[5-(trifluoromethyl)pyridin-2-yl]phenyl}methanamine
Molecular Weight (g/mol): 252.24
Molecular Formula: C13H11F3N2
InChi Key: XQAJYHFQYZEWSE-UHFFFAOYSA-N
SMILES: NCC1=CC=C(C=C1)C1=NC=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results