CAS RN 906352-79-2
IUPAC Name:
[6-(oxan-4-yloxy)pyridin-3-yl]methanol
Synonyms:
[6-(oxan-4-yloxy)pyridin-3-yl]methanol
Molecular Weight (g/mol):
209.25
Molecular Formula:
C11H15NO3
InChi Key:
UTEOAQJLPJOMJS-UHFFFAOYSA-N
SMILES:
OCC1=CN=C(OC2CCOCC2)C=C1