accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNO

CAS RN 906352-82-7

IUPAC Name: 1-(6-methylpyrazin-2-yl)piperidine-4-carbaldehyde
Synonyms: 1-(6-methylpyrazin-2-yl)piperidine-4-carbaldehyde
Molecular Weight (g/mol): 205.26
Molecular Formula: C11H15N3O
InChi Key: KWDJDKVUDZLNOD-UHFFFAOYSA-N
SMILES: CC1=CN=CC(=N1)N1CCC(CC1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results