CAS RN 906480-09-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Angiopep-2-Cys
Binds to LRP1 and facilitates receptor-mediated transcytosis of cargo across the BBB