accessibility menu, dialog, popup

UserName

Viewing profile as user:
BSFFFK

CAS RN 906674-55-3

IUPAC Name: potassium trifluoro(thiophen-2-yl)boranuide
Synonyms: potassium trifluoro(thiophen-2-yl)boranuide
Molecular Weight (g/mol): 190.03
Molecular Formula: C4H3BF3KS
InChi Key: LXOCUYMZBWNQQS-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results