Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium 2-Thienyltrifluoroborate 98.0+%, TCI America™
SDP

Catalog No. P18285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18285G 5 g
P18281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18285G Supplier TCI America Supplier No. P18285G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 906674-55-3
Molecular Formula C4H3BF3KS
Molecular Weight (g/mol) 190.032
MDL Number MFCD04112719
InChI Key LXOCUYMZBWNQQS-UHFFFAOYSA-N
PubChem CID 44630027
IUPAC Name potassium;trifluoro(thiophen-2-yl)boranuide
SMILES [B-](C1=CC=CS1)(F)(F)F.[K+]

Specifications

Melting Point 253°C
Color White-Yellow
Quantity 5 g
Formula Weight 190.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium 2-Thienyltrifluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.