CAS RN 9068-54-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MP Biomedicals, Inc Phosphodiesterase II, MP Biomedicals™
CAS: 9068-54-6 MDL Number: MFCD00131884 Synonym: Spleen phosphodiesterase,Spleen exonuclease