Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphodiesterase II, MP Biomedicals™

Catalog No. ICN10097701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10097701 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10097701 Supplier MP Biomedicals, Inc Supplier No. 0210097701

Chemical Identifiers

CAS 9068-54-6
MDL Number MFCD00131884
Synonym Spleen phosphodiesterase, Spleen exonuclease

Specifications

Color White
Quantity 1 Vial
Physical Form Solid
Chemical Name or Material L-Pipecolic Acid
Recommended Storage Store at -20°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.