accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOH

CAS RN 90721-27-0

IUPAC Name: 1-benzofuran-5-carboxylic acid
Synonyms: 1-benzofuran-5-carboxylic acid
Molecular Weight (g/mol): 162.14
Molecular Formula: C9H6O3
InChi Key: GTWXSZIQNTUNKR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2OC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results