Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzofuran-5-carboxylic acid, Thermo Scientific™

Catalog No. CC07101DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC07101DE 5 g Amber Glass Bottle
CC07101CB 250 mg Amber Glass Bottle
CC07101DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC07101DE Supplier Thermo Scientific Chemicals Supplier No. CC07101DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90721-27-0
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD01006742
InChI Key GTWXSZIQNTUNKR-UHFFFAOYSA-N
Synonym benzofuran-5-carboxylic acid, 5-benzofurancarboxylic acid, benzo b furan-5-carboxylic acid, 1-benzofunan-5-carboxylic acid, benzofurane-5-carboxylic acid, 4ctj, pubchem7018, 5-benzofuran carboxylic, rarechem al be 1369, 5-benzofurancarboxylicacid
PubChem CID 595656
IUPAC Name 1-benzofuran-5-carboxylic acid
SMILES OC(=O)C1=CC=C2OC=CC2=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 162.14
Percent Purity 97%
Chemical Name or Material 1-Benzofuran-5-carboxylic acid
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.