accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3H3COHOH2N

CAS RN 9073-63-6

IUPAC Name: 2-aminoacetic acid; bis(butane)
Synonyms: bis(butane); glycine
Molecular Weight (g/mol): 191.32
Molecular Formula: C10H25NO2
InChi Key: LJCNDNBULVLKSG-UHFFFAOYSA-N
SMILES: CCCC.CCCC.NCC(O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results