Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol Oxidase, MP Biomedicals™

Catalog No. MP021901552 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021901552 250 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021901552 Supplier MP Biomedicals, Inc Supplier No. 02190155.2
Add to Cart
Add to Cart

Plays a major role in the metabolism of methanol, resulting in the formation of formaldehyde. Alcohol Oxidase, MP Biomedicals is used for qualitative and quantitative determination of methanol or ethanol (blood alcohol) by colorimetric procedures, and can

Alcohol oxidase has been detected in several strains of yeast (such as Candida, Pichia and Hansenula) that utilize methanol as a sole carbon and energy source. Some of these enzymes have been isolated and partially characterized. Monomers are synthesized in the cytosol and assembled into octomers in the peroxisome. Octomerization is thought to be chaparone mediated.

Alcohol oxidase is of interest for the study of protein translocation into peroxisomes. It catalyzes the oxidation of an alcohol to an aldehyde and hydrogen peroxide.

  • Catalyzes the oxidation of short-chain, primary, aliphatic alcohols to their respective aldehydes (it has the highest affinity for methanol)
  • Involved in the metabolism of methanol, which is important for yeasts that utilize methanol as their sole energy source
  • Primarily found in the peroxisome, but may be present in the cytoplasm as well
  • One unit will oxidize 1.0 μmole of methanol to formaldehyde per minute at pH 7.5 at 25°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9073-63-6
Molecular Formula C10H25NO2
Molecular Weight (g/mol) 191.315
InChI Key LJCNDNBULVLKSG-UHFFFAOYSA-N
Synonym oxidase, alcohol, bis butane ; glycine
PubChem CID 73555276
IUPAC Name 2-aminoacetic acid;butane
SMILES CCCC.CCCC.C(C(=O)O)N

Specifications

Quantity 250 U
Formula Weight about 630,000g/mol
Physical Form Frozen Liquid
Recommended Storage Freeze at –20°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.