CAS RN 908142-03-0
IUPAC Name:
[3-(hydroxymethyl)-4-methoxyphenyl]boronic acid
Synonyms:
3-(hydroxymethyl)-4-methoxyphenylboronic acid
Molecular Weight (g/mol):
181.98
Molecular Formula:
C8H11BO4
InChi Key:
DOQUCBSZQOPLGT-UHFFFAOYSA-N
SMILES:
COC1=C(CO)C=C(C=C1)B(O)O