accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 908847-01-8

FOOOHONHHN(S)

CAS RN 908847-01-8

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(6-fluoro-1H-indol-3-yl)propanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(6-fluoro-1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol): 444.46
Molecular Formula: C26H21FN2O4
InChi Key: ZNQAUDMHOJUTJL-DEOSSOPVSA-N
SMILES: OC(=O)[C@H](CC1=CNC2=CC(F)=CC=C12)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)