accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(S)NH3

CAS RN 90891-21-7

IUPAC Name: (2S)-1-ethoxy-1-oxo-4-phenylbutan-2-aminium
Synonyms: (2S)-1-ethoxy-1-oxo-4-phenylbutan-2-aminium
Molecular Weight (g/mol): 208.28
Molecular Formula: C12H18NO2
InChi Key: WFLQXECQLHZKMV-NSHDSACASA-O
SMILES: CCOC(=O)[C@@H]([NH3+])CCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)