accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 909036-46-0

IUPAC Name: 2-chloro-3-iodopyridin-4-amine
Synonyms: 2-chloro-3-iodopyridin-4-amine
Molecular Weight (g/mol): 254.46
Molecular Formula: C5H4ClIN2
InChi Key: BYVMKCHHWASQFN-UHFFFAOYSA-N
SMILES: NC1=C(I)C(Cl)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results