Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-chloro-3-iodopyridine, 95%, Thermo Scientific™

Catalog No. AAH3313603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3313603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3313603 Supplier Thermo Scientific Chemicals Supplier No. H3313603
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 909036-46-0
Molecular Formula C5H4ClIN2
Molecular Weight (g/mol) 254.46
MDL Number MFCD09763660
InChI Key BYVMKCHHWASQFN-UHFFFAOYSA-N
Synonym 2-chloro-3-iodo-4-pyridinamine, 2-chloro-3-iodo-4-aminopyridine, 4-amino-2-chloro-3-iodopyridine, pubchem22625, 2-chloro-3-iodo-4-pyridylamine, 2-chloro-3-iodo-pyridin-4-amine, 2-chloro-3-iodopyridin-4-ylamine, 2-chloro-3-iodo-pyridin-4-ylamine, 4-pyridinamine, 2-chloro-3-iodo
PubChem CID 24229208
IUPAC Name 2-chloro-3-iodopyridin-4-amine
SMILES NC1=C(I)C(Cl)=NC=C1

Specifications

Melting Point 116°C to 117°C
Color Orange-Pink to White
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 254.46
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Amino-2-chloro-3-iodopyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.