CAS RN 909115-33-9
IUPAC Name:
(4S)-3-[(2S)-5-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
Synonyms:
(4S)-3-[(2S)-5-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-oxopentanoyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
Molecular Weight (g/mol):
552.62
Molecular Formula:
C30H36N2O8
InChi Key:
DSGXIRQTDFOKPQ-ZEQRLZLVSA-N
SMILES:
CC(C)(C)OC(=O)CC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(=O)N1[C@@H](COC1(C)C)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-Glu OtBu -Ser psiMe.Mepro, Advanced ChemTech
CAS: 909115-33-9 Molecular Formula: C30H36N2O8 Molecular Weight (g/mol): 552.62 MDL Number: MFCD18427719 InChI Key: DSGXIRQTDFOKPQ-ZEQRLZLVSA-N IUPAC Name: (4S)-3-[(2S)-5-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid SMILES: CC(C)(C)OC(=O)CC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(=O)N1[C@@H](COC1(C)C)C(O)=O