accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90914-81-1

IUPAC Name: N-(4-bromo-3-methylphenyl)acetamide
Synonyms: 3-methyl-4-bromoacetanilide
Molecular Weight (g/mol): 228.09
Molecular Formula: C9H10BrNO
InChi Key: BYZHUFNLXFFINU-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC(C)=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results