Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Bromo-3'-methylacetanilide 98.0+%, TCI America™
SDP

Catalog No. B31425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31425G 5 g
B314225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31425G Supplier TCI America Supplier No. B31425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90914-81-1
Molecular Formula C9H10BrNO
Molecular Weight (g/mol) 228.089
MDL Number MFCD00040822
InChI Key BYZHUFNLXFFINU-UHFFFAOYSA-N
Synonym n-4-bromo-3-methylphenyl acetamide, 4'-bromo-3'-methylacetanilide, 4-bromo-3-methylacetanilide, n-acetyl-4-bromo-m-toluidine, 3-methyl-4-bromoacetanilide, n-acetyl 4-bromo-3-methylaniline, n-4-bromo-3-methylphenyl-acetamide, maybridge1_005107, acmc-209r7e
PubChem CID 738336
IUPAC Name N-(4-bromo-3-methylphenyl)acetamide
SMILES CC1=C(C=CC(=C1)NC(=O)C)Br

Specifications

Melting Point 103°C
Quantity 5 g
Formula Weight 228.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromo-3′-methylacetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.