accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90916-29-3

IUPAC Name: prop-2-en-1-yl N-(2-nitrobenzenesulfonyl)carbamate
Synonyms: prop-2-en-1-yl N-(2-nitrobenzenesulfonyl)carbamate
Molecular Weight (g/mol): 286.26
Molecular Formula: C10H10N2O6S
InChi Key: CRGOQFIAULBODK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1S(=O)(=O)NC(=O)OCC=C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results