Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Allyloxycarbonyl-2-nitrobenzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. A16325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16325G Supplier TCI America Supplier No. A16325G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90916-29-3
Molecular Formula C10H10N2O6S
Molecular Weight (g/mol) 286.26
MDL Number MFCD06797039
InChI Key CRGOQFIAULBODK-UHFFFAOYSA-N
Synonym N-Alloc-2-nitrobenzenesulfonamide
PubChem CID 11033445
IUPAC Name prop-2-en-1-yl N-(2-nitrobenzenesulfonyl)carbamate
SMILES [O-][N+](=O)C1=CC=CC=C1S(=O)(=O)NC(=O)OCC=C

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 5 g
Formula Weight 286.26
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Allyloxycarbonyl-2-nitrobenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.