accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 909254-56-4

IUPAC Name: 6-nitro-19,40-dioxa-2,10,16,43,48,49-hexaazaoctacyclo[42.3.1.1⁴,⁸.1¹¹,¹⁵.0²⁰,²⁹.0²³,²⁸.0³⁰,³⁹.0³¹,³⁶]pentaconta-1(48),4,6,8(50),11(49),12,14,20,22,24,26,28,30,32,34,36,38,44,46-nonadecaene-3,9,17,42-tetrone
Synonyms: 6-nitro-19,40-dioxa-2,10,16,43,48,49-hexaazaoctacyclo[42.3.1.1⁴,⁸.1¹¹,¹⁵.0²⁰,²⁹.0²³,²⁸.0³⁰,³⁹.0³¹,³⁶]pentaconta-1(48),4,6,8(50),11(49),12,14,20,22,24,26,28,30,32,34,36,38,44,46-nonadecaene-3,9,17,42-tetrone
Molecular Weight (g/mol): 759.74
Molecular Formula: C42H29N7O8
InChi Key: AFQYUGXRHMUXOZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC2=CC(=C1)C(=O)NC1=NC(NC(=O)COC3=CC=C4C=CC=CC4=C3C3=C4C=CC=CC4=CC=C3OCC(=O)NC3=CC=CC(NC2=O)=N3)=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results