Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chirabite-AR, TCI America™
SDP

Catalog No. C2184100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2184100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2184100MG Supplier TCI America Supplier No. C2184100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 909254-56-4
Molecular Formula C42H29N7O8
Molecular Weight (g/mol) 759.74
MDL Number MFCD09038511
InChI Key AFQYUGXRHMUXOZ-UHFFFAOYSA-N
Synonym (R)-2,2′C-[5-Nitroisophthalamidobis(2,6-pyridylenecarbamoylmethoxy)]-1,1′C-binaphthyl
PubChem CID 25112234
IUPAC Name 6-nitro-19,40-dioxa-2,10,16,43,48,49-hexaazaoctacyclo[42.3.1.1⁴,⁸.1¹¹,¹⁵.0²⁰,²⁹.0²³,²⁸.0³⁰,³⁹.0³¹,³⁶]pentaconta-1(48),4,6,8(50),11(49),12,14,20,22,24,26,28,30,32,34,36,38,44,46-nonadecaene-3,9,17,42-tetrone
SMILES [O-][N+](=O)C1=CC2=CC(=C1)C(=O)NC1=NC(NC(=O)COC3=CC=C4C=CC=CC4=C3C3=C4C=CC=CC4=CC=C3OCC(=O)NC3=CC=CC(NC2=O)=N3)=CC=C1

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 759.74
Physical Form Crystalline Powder
Chemical Name or Material Chirabite-AR
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.