accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOBOBO

CAS RN 909407-14-3

IUPAC Name: tris({5-phenyl-[1,1'-biphenyl]-3-yl})-1,3,5,2,4,6-trioxatriborinane
Synonyms: tris({5-phenyl-[1,1'-biphenyl]-3-yl})-1,3,5,2,4,6-trioxatriborinane
Molecular Weight (g/mol): 768.33
Molecular Formula: C54H39B3O3
InChi Key: MWZSDMFLGCWWSC-UHFFFAOYSA-N
SMILES: O1B(OB(OB1C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results