Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tris(m-terphenyl-5'-yl)boroxin 95.0+%, TCI America™
SDP

Catalog No. T1887100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1887100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1887100MG Supplier TCI America Supplier No. T1887100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 909407-14-3
Molecular Formula C54H39B3O3
Molecular Weight (g/mol) 768.33
MDL Number MFCD04038427
InChI Key MWZSDMFLGCWWSC-UHFFFAOYSA-N
Synonym (m-Terphenyl-5′C-yl)boronic Anhydride, Tris(m-terphenyl-5′C-yl)cyclotriboroxane
PubChem CID 44630523
IUPAC Name tris({5-phenyl-[1,1'-biphenyl]-3-yl})-1,3,5,2,4,6-trioxatriborinane
SMILES O1B(OB(OB1C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 100 mg
Formula Weight 768.33
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tris(m-terphenyl-5′-yl)boroxin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.