accessibility menu, dialog, popup

UserName

Viewing profile as user:
BCH3FHOOH

CAS RN 909709-42-8

IUPAC Name: {3-fluoro-4'-propyl-[1,1'-biphenyl]-4-yl}boronic acid
Synonyms: 3-fluoro-4'-propyl-[1,1'-biphenyl]-4-ylboronic acid
Molecular Weight (g/mol): 258.1
Molecular Formula: C15H16BFO2
InChi Key: XZGCBSYWWZAXRC-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(C=C1)C1=CC(F)=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results