Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4'-propyl-4-biphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F09425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0942-5G 5 g
F09421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0942-5G Supplier TCI America Supplier No. F09425G
Add to Cart
Add to Cart

Specifications

CAS 909709-42-8
Color White-Yellow
Molecular Formula C15H16BFO2
Quantity 5 g
MDL Number MFCD10687198
Synonym 3-fluoro-4 '-propyl-1,1 '-biphenyl-4-yl boronic acid, 4 '-propyl-3-fluorobiphenyl-4-boronic acid, 4-propyl-3-fluorobiphenyl-4-boronic acid, 3-fluoro-4 '-propyl-biphenylboronic acid, 3-fluoro-4 '-propyl-4-biphenylboronic acid
InChI Key XZGCBSYWWZAXRC-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)C2=CC=C(C=C2)CCC)F)(O)O
IUPAC Name [2-fluoro-4-(4-propylphenyl)phenyl]boronic acid
Molecular Weight (g/mol) 258.099
PubChem CID 53402576
Formula Weight 258.10
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.